(4-Chloro-3-pyridinyl)methanol
Lieferant: Apollo Scientific
|
Achtung
|
OR13855-1G
APOSOR13855-1GEA
87
EUR
APOSOR13855-1G
(4-Chloro-3-pyridinyl)methanol
(4-Chloro-3-pyridinyl)methanol
Formel:
C₆H₆ClNO Schmelzpunkt: 94…96 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09743993 CAS-Nummer: 189449-41-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...