2-Amino-4-methoxypyridin

Lieferant: Apollo Scientific
Achtung

OR13856-5G OR13856-25G
APOSOR13856-5GEA 39.3 EUR
APOSOR13856-5G APOSOR13856-25G
2-Amino-4-methoxypyridin
2-Amino-4-methoxypyridine
Formel: C₆H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD07437849
CAS-Nummer: 10201-73-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR