4-(1-Pyrrolidinyl)benzaldehyd

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(1-Pyrrolidino)benzaldehyde, 4-(1-Pyrrolidinyl)benzaldehyde

OR1386-1G
APOSOR1386-1GEA 21.9 EUR
APOSOR1386-1G
4-(1-Pyrrolidinyl)benzaldehyd
1-(4-Formylphenyl)pyrrolidine
Formel: C₁₁H₁₃NO
Molecular Weight: 175,23 g/mol
Schmelzpunkt: 81…85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00015469
CAS-Nummer: 51980-54-2
EINECS: 257-570-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR