Ethyl-4-hydroxymandelate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 4-hydroxymandelate

OR13893-250MG
APOSOR13893-250MGEA 21.9 EUR
APOSOR13893-250MG
Ethyl-4-hydroxymandelate
Ethyl-4-hydroxymandelate
Formel: C₁₀H₁₂O₄
Molecular Weight: 196.20 g/mol
Schmelzpunkt: 128…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00020180
CAS-Nummer: 68758-68-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR