2-Thien-2-ylbenzaldehyde 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Thiophen-2-yl)benzaldehyde

OR1392-1G
APOSOR1392-1GEA 212 EUR
APOSOR1392-1G
2-Thien-2-ylbenzaldehyde 97%
2-Thien-2-ylbenzaldehyde
Formel: C₁₁H₈OS
Molecular Weight: 188,25 g/mol
Siedepunkt: 170 °C/0,5mmHg
Lagertemperatur: Kühlschrank
MDL: MFCD04039148
CAS-Nummer: 99902-07-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR