5-Methyl-1-phenyl-1H-pyrazole-4-boronic acid, pinacol ester 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Methyl-1-phenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 2-(5-Methyl-1-phenyl-1H-pyrazol-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR1395-250MG
APOSOR1395-250MGEA 112 EUR
APOSOR1395-250MG
5-Methyl-1-phenyl-1H-pyrazole-4-boronic acid, pinacol ester 97%
5-Methyl-1-phenyl-1H-pyrazole-4-boronic acid, pinacol ester
Formel: C₁₆H₂₁BN₂O₂
Molecular Weight: 284,161 g/mol
Schmelzpunkt: 110…112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06797463
CAS-Nummer: 849776-88-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR