3-(Morpholinomethyl)phenylboronsäurepinakolester 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR1396-1G
OR1396-250MG
APOSOR1396-1GEA
0
EUR
APOSOR1396-1G
APOSOR1396-250MG
3-(Morpholinomethyl)phenylboronsäurepinakolester 97%
3-(Morpholinomethyl)phenylboronsäurepinakolester
Formel:
C₁₇H₂₆BNO₃ Schmelzpunkt: 50…53 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797489 CAS-Nummer: 364794-80-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...