4-Benzyl-2-(hydroxymethyl)piperazine 95+%

Lieferant: Apollo Scientific

Synonyme: (4-Benzylpiperazin-2-yl)methanol

OR13971-250MG
APOSOR13971-250MGEA 102 EUR
APOSOR13971-250MG
4-Benzyl-2-(hydroxymethyl)piperazine 95+%
4-Benzyl-2-(hydroxymethyl)piperazine
Formel: C₁₂H₁₈N₂O
Molecular Weight: 206,284 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03001699
CAS-Nummer: 85817-34-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR