1-(4-Methoxyphenyl)-N-methylmethanamin

Lieferant: Apollo Scientific
Gefahr

OR13981-250MG
APOSOR13981-250MGEA 21.9 EUR
APOSOR13981-250MG
1-(4-Methoxyphenyl)-N-methylmethanamin
4-Methoxy-N-methylbenzylamine
Formel: C₉H₁₃NO
Molecular Weight: 151,21 g/mol
Siedepunkt: 88…90 °C/1mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD04115407
CAS-Nummer: 702-24-9
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR