4-Cyan-4'-heptylbiphenyl 98%

Lieferant: Apollo Scientific

Synonyme: 4-Cyano-4'-n-heptylbiphenyl, 4'-Heptyl-[1,1'-biphenyl]-4-carbonitrile

OR13990-25G
APOSOR13990-25GEA 95.7 EUR
APOSOR13990-25G
4-Cyan-4'-heptylbiphenyl 98%
4-Cyan-4'-heptylbiphenyl
Formel: C₂₀H₂₃N
Molecular Weight: 277.41 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00074915
CAS-Nummer: 41122-71-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR