4-Butyl-4'-cyanbiphenyl 98%

Lieferant: Apollo Scientific

Synonyme: 4'-Butyl-[1,1'-biphenyl]-4-carbonitrile

OR13992-25G
APOSOR13992-25GEA 157 EUR
APOSOR13992-25G
4-Butyl-4'-cyanbiphenyl 98%
4-Butyl-4'-cyanbiphenyl
Formel: C₁₇H₁₇N
Molecular Weight: 235,33 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00506009
CAS-Nummer: 52709-83-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR