4'-Hydroxybiphenyl-4-carbonsäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4'-Hydroxy-4-biphenylcarboxylic acid, 4'-Hydroxy-[1,1'-biphenyl]-4-carboxylic acid
OR13999-25G
OR13999-5G
OR13999-100G
APOSOR13999-25GEA
60.9
EUR
APOSOR13999-25G
APOSOR13999-5G
APOSOR13999-100G
4'-Hydroxybiphenyl-4-carbonsäure 95%
4'-Hydroxybiphenyl-4-carbonsäure
Formel:
C₁₃H₁₀O₃ Molecular Weight: 214.22 g/mol Schmelzpunkt: 295 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059078 CAS-Nummer: 58574-03-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...