2-Hydroxyquinoline-3-carbaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 2(1H)-Quinolinone-3-carboxaldehyde, 2-Oxo-1,2-dihydro-3-quinolinecarbaldehyde

OR14001-1G OR14001-5G
APOSOR14001-1GEA 105 EUR
APOSOR14001-1G APOSOR14001-5G
2-Hydroxyquinoline-3-carbaldehyde
2-Hydroxyquinoline-3-carbaldehyde
Formel: C₁₀H₇NO₂
Molecular Weight: 173.171 g/mol
Schmelzpunkt: >250 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00563443
CAS-Nummer: 91301-03-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR