2-Hydroxyquinoline-3-carbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2(1H)-Quinolinone-3-carboxaldehyde, 2-Oxo-1,2-dihydro-3-quinolinecarbaldehyde
OR14001-1G
OR14001-5G
APOSOR14001-1GEA
105
EUR
APOSOR14001-1G
APOSOR14001-5G
2-Hydroxyquinoline-3-carbaldehyde
2-Hydroxyquinoline-3-carbaldehyde
Formel:
C₁₀H₇NO₂ Molecular Weight: 173.171 g/mol Schmelzpunkt: >250 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00563443 CAS-Nummer: 91301-03-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...