3-(Hydroxymethyl)-2(1H)-quinolinone 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Hydroxymethyl)quinolin-2(1H)-one

OR14005-1G
APOSOR14005-1GEA 336 EUR
APOSOR14005-1G
3-(Hydroxymethyl)-2(1H)-quinolinone 95%
3-(Hydroxymethyl)-2(1H)-quinolinone
Formel: C₁₀H₉NO₂
Molecular Weight: 175,19 g/mol
Schmelzpunkt: 200 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06200995
CAS-Nummer: 90097-45-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR