5-Iod-2-(4-morpholino)pyridin

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(5-Iodo-2-pyridinyl)morpholine, 4-(5-Iodo-2-pyridyl)morpholine

OR14007-250MG
APOSOR14007-250MGEA 23.2 EUR
APOSOR14007-250MG
5-Iod-2-(4-morpholino)pyridin
5-Iod-2-(4-morpholino)pyridin
Formel: C₉H₁₁IN₂O
Molecular Weight: 290,11 g/mol
Schmelzpunkt: 129…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04039866
CAS-Nummer: 470463-42-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR