6-tert-Butyl-3-ethyl-2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate
Lieferant: Apollo Scientific
Synonyme:
Ethyl 2-amino-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate, N-BOC protected, Ethyl 6-Boc-2-amino-4,7-dihydro-5H-thieno[2,3-c]pyridine-3-carboxylate
OR14010-1G
APOSOR14010-1GEA
29
EUR
APOSOR14010-1G
6-tert-Butyl-3-ethyl-2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate
6-tert-Butyl-3-ethyl-2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate
Formel:
C₁₅H₂₂N₂O₄S Molecular Weight: 326,42 g/mol Schmelzpunkt: 153…154 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05664039 CAS-Nummer: 193537-14-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...