6-Pyrrolidin-1-ylnicotinaldehyde

Lieferant: Apollo Scientific
Achtung

OR14012-5G OR14012-1G
APOSOR14012-5GEA 0 EUR
APOSOR14012-5G APOSOR14012-1G
6-Pyrrolidin-1-ylnicotinaldehyde
6-Pyrrolidin-1-ylnicotinaldehyde
Formel: C₁₀H₁₂N₂O
Schmelzpunkt: 88…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06200881
CAS-Nummer: 261715-39-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR