5-Methylbenzo[b]thiophene-2-sulphonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR14014-1G
OR14014-250MG
APOSOR14014-1GEA
377
EUR
APOSOR14014-1G
APOSOR14014-250MG
5-Methylbenzo[b]thiophene-2-sulphonyl chloride
5-Methylbenzo[b]thiophene-2-sulphonyl chloride
Formel:
C₉H₇ClO₂S₂ Molecular Weight: 246.734 g/mol Schmelzpunkt: 83 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06200880 CAS-Nummer: 90273-30-6 UN: 2923 ADR: 8+6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...