4-Brom-2-benzo[b]thiophencarbonsäure

Lieferant: Apollo Scientific

OR14031-1G
APOSOR14031-1GEA 55.1 EUR
APOSOR14031-1G
4-Brom-2-benzo[b]thiophencarbonsäure
4-Brom-2-benzo[b]thiophencarbonsäure
Formel: C₉H₅BrO₂S
Molecular Weight: 257.1038 g/mol
Schmelzpunkt: 265 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03407342
CAS-Nummer: 5194-37-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR