2-Bromo-1-(5-chloro-2-thienyl)-1-ethanone

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromo-1-(5-chlorothiophen-2-yl)ethanone, 2-(Bromoacetyl)-5-chlorothiophene

OR14033-1G
APOSOR14033-1GEA 21.9 EUR
APOSOR14033-1G
2-Bromo-1-(5-chloro-2-thienyl)-1-ethanone
2-Bromo-1-(5-chloro-2-thienyl)-1-ethanone
Formel: C₆H₄BrClOS
Molecular Weight: 239.52 g/mol
Schmelzpunkt: 73…75 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00829820
CAS-Nummer: 57731-17-6
UN: 1759
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR