4-Chlor-2-benzo[b]thiophencarbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR14042-1G
OR14042-5G
APOSOR14042-1GEA
46.4
EUR
APOSOR14042-1G
APOSOR14042-5G
4-Chlor-2-benzo[b]thiophencarbonsäure
4-Chlor-2-benzo[b]thiophencarbonsäure
Formel:
C₉H₅ClO₂S Schmelzpunkt: 257…259 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06655155 CAS-Nummer: 23967-57-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...