4-Chlorphenacylthiocyanat

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Chlorophenyl)-2-thiocyanatoethanone, 2-(4-Chlorophenyl)-2-oxoethyl thiocyanate

OR14046-1G
APOSOR14046-1GEA 34.8 EUR
APOSOR14046-1G
4-Chlorphenacylthiocyanat
4-Chlorphenacylthiocyanat
Formel: C₉H₆ClNOS
Molecular Weight: 211,668 g/mol
Schmelzpunkt: 136…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00019771
CAS-Nummer: 19339-59-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR