2-(3-Chloro-2-thienyl)-2-oxoethyl thiocyanate
Lieferant: Apollo Scientific
OR14048-500MG
APOSOR14048-500MGEA
560
EUR
APOSOR14048-500MG
2-(3-Chloro-2-thienyl)-2-oxoethyl thiocyanate
2-(3-Chloro-2-thienyl)-2-oxoethyl thiocyanate
Formel:
C₇H₄ClNOS₂ Molecular Weight: 217,7 g/mol Schmelzpunkt: 76…78 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09027129 CAS-Nummer: 937602-35-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...