Methyl-6-chlor-2-benzo[b]thiophencarboxylat

Lieferant: Apollo Scientific

OR14067-1G
APOSOR14067-1GEA 0 EUR
APOSOR14067-1G
Methyl-6-chlor-2-benzo[b]thiophencarboxylat
Methyl-6-chlor-2-benzo[b]thiophencarboxylat
Formel: C₁₀H₇ClO₂S
Schmelzpunkt: 107…109 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027103
CAS-Nummer: 104795-85-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR