Methyl-5-chlor-2-benzo[b]thiophencarboxylat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 5-chlorobenzo[b]thiophene-2-carboxylate
OR14068-1G
APOSOR14068-1GEA
56.7
EUR
APOSOR14068-1G
Methyl-5-chlor-2-benzo[b]thiophencarboxylat
Methyl-5-chlor-2-benzo[b]thiophencarboxylat
Formel:
C₁₀H₇ClO₂S Schmelzpunkt: 113…115 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09027104 CAS-Nummer: 35212-96-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...