Methyl-7-chlor-2-benzo[b]thiophencarboxylat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 7-chlorobenzo[b]thiophene-2-carboxylate
OR14069-1G
APOSOR14069-1GEA
68.3
EUR
APOSOR14069-1G
Methyl-7-chlor-2-benzo[b]thiophencarboxylat
Methyl-7-chlor-2-benzo[b]thiophencarboxylat
Formel:
C₁₀H₇ClO₂S Molecular Weight: 226,6794 g/mol Schmelzpunkt: 86…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09027105 CAS-Nummer: 550998-56-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...