2-(Allyloxy)benzaldehyd

Lieferant: Apollo Scientific

OR14086-25G
APOSOR14086-25GEA 116 EUR
APOSOR14086-25G
2-(Allyloxy)benzaldehyd
2-(Allyloxy)benzaldehyd
Formel: C₁₀H₁₀O₂
Siedepunkt: 92…94 °C/0,04mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00014130
CAS-Nummer: 28752-82-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR