Thiomorpholine-3-carboxylic acid, N-BOC protected
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(tert-Butoxycarbonyl)-3-carboxythiomorpholine, 4-(tert-Butoxycarbonyl)thiomorpholine-3-carboxylic acid
OR1411-250MG
APOSOR1411-250MGEA
59.6
EUR
APOSOR1411-250MG
Thiomorpholine-3-carboxylic acid, N-BOC protected
Thiomorpholine-3-carboxylic acid, N-BOC protected
Formel:
C₁₀H₁₇NO₄S Schmelzpunkt: 145…147 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02683067 CAS-Nummer: 128453-98-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...