2-(4-Methoxybenzoyl)benzenecarboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-Methoxybenzoyl)benzoic acid

OR14127-1G
APOSOR14127-1GEA 68.3 EUR
APOSOR14127-1G
2-(4-Methoxybenzoyl)benzenecarboxylic acid 95%
2-(4-Methoxybenzoyl)benzenecarboxylic acid
Formel: C₁₅H₁₂O₄
Molecular Weight: 256,26 g/mol
Schmelzpunkt: 147…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00278217
CAS-Nummer: 1151-15-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR