2-(4-Toluoyl)benzoesäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(4-Methylbenzoyl)benzenecarboxylic acid
OR14136-1G
APOSOR14136-1GEA
21.9
EUR
APOSOR14136-1G
2-(4-Toluoyl)benzoesäure
2-(4-Toluoyl)benzoesäure
Formel:
C₁₅H₁₂O₃ Molecular Weight: 240,26 g/mol Schmelzpunkt: 137…138 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00020287 CAS-Nummer: 85-55-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...