Methyl-3-bromo-4-hydroxy-5-nitrobenzenecarboxylate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 3-bromo-4-hydroxy-5-nitrobenzoate
OR14142-1G
APOSOR14142-1GEA
42.2
EUR
APOSOR14142-1G
Methyl-3-bromo-4-hydroxy-5-nitrobenzenecarboxylate 95%
Methyl-3-bromo-4-hydroxy-5-nitrobenzenecarboxylate
Formel:
C₈H₆BrNO₅ Molecular Weight: 276,05 g/mol Schmelzpunkt: 131…133 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08689690 CAS-Nummer: 40258-72-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...