1-Methyl-5-nitro-1H-indazol

Lieferant: Apollo Scientific
Achtung

OR14146-1G
APOSOR14146-1GEA 21.9 EUR
APOSOR14146-1G
1-Methyl-5-nitro-1H-indazol
1-Methyl-5-nitro-1H-indazole
Formel: C₈H₇N₃O₂
Molecular Weight: 177,1600 g/mol
Schmelzpunkt: 159…161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01318163
CAS-Nummer: 5228-49-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR