8-Amino-7-methylquinoline
Lieferant: Apollo Scientific
|
Achtung
|
OR14149-1G
APOSOR14149-1GEA
0
EUR
APOSOR14149-1G
8-Amino-7-methylquinoline
8-Amino-7-methylquinoline
Formel:
C₁₀H₁₀N₂ Schmelzpunkt: 46…48 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00168259 CAS-Nummer: 5470-82-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...