3-(Isopropoxycarbonyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Isopropoxycarbonyl)benzeneboronic acid

OR14155-250MG
APOSOR14155-250MGEA 21.9 EUR
APOSOR14155-250MG
3-(Isopropoxycarbonyl)phenylboronsäure 98%
3-(Isopropoxycarbonyl)phenylboronsäure
Formel: C₁₀H₁₃BO₄
Molecular Weight: 208,02 g/mol
Schmelzpunkt: 153…154 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093047
CAS-Nummer: 342002-80-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR