1-(3-Bromophenyl)-1H-pyrazole 97%

Lieferant: Apollo Scientific
Achtung

OR1418-1G
APOSOR1418-1GEA 23.2 EUR
APOSOR1418-1G
1-(3-Bromophenyl)-1H-pyrazole 97%
1-(3-Bromophenyl)-1H-pyrazole
Formel: C₉H₇BrN₂
Molecular Weight: 223,0693 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06797472
CAS-Nummer: 294877-33-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR