cis-3-Azabicyclo[3.1.0]hexane-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
cis-2-Carboxy-3-azabicyclo[3.1.0]hexane
OR14181-250MG
APOSOR14181-250MGEA
71.2
EUR
APOSOR14181-250MG
cis-3-Azabicyclo[3.1.0]hexane-2-carboxylic acid
cis-3-Azabicyclo[3.1.0]hexane-2-carboxylic acid
Formel:
C₆H₉NO₂ Schmelzpunkt: 252…256 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03093875 CAS-Nummer: 1279569-17-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...