5-Amino-1-(3,4-dichlorophenyl)-1H-pyrazole-4-carbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR14183-1G
APOSOR14183-1GEA
34.8
EUR
APOSOR14183-1G
5-Amino-1-(3,4-dichlorophenyl)-1H-pyrazole-4-carbonitrile
5-Amino-1-(3,4-dichlorophenyl)-1H-pyrazole-4-carbonitrile
Formel:
C₁₀H₆Cl₂N₄ Schmelzpunkt: 170 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00128311 CAS-Nummer: 58791-78-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...