5-Amino-1-(4-methylphenyl)-1H-pyrazole-4-carbonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-1-(p-tolyl)-1H-pyrazole-4-carbonitrile, 5-Amino-1-(4-methylphenyl)pyrazole-4-carbonitrile, 5-Amino-1-(p-tolyl)pyrazole-4-carbonitrile

OR14185-1G
APOSOR14185-1GEA 43.5 EUR
APOSOR14185-1G
5-Amino-1-(4-methylphenyl)-1H-pyrazole-4-carbonitrile
5-Amino-1-(4-methylphenyl)-1H-pyrazole-4-carbonitrile
Formel: C₁₁H₁₀N₄
Molecular Weight: 198,23 g/mol
Schmelzpunkt: 116…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052944
CAS-Nummer: 103646-82-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR