3-Bromoimidazo[1,2-a]pyrimidine 98%

Lieferant: Apollo Scientific
Achtung

OR14190-1G
APOSOR14190-1GEA 29 EUR
APOSOR14190-1G
3-Bromoimidazo[1,2-a]pyrimidine 98%
3-Bromoimidazo[1,2-a]pyrimidine
Formel: C6H4BrN3
Molecular Weight: 198,0200 g/mol
Schmelzpunkt: 158…160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06200962
CAS-Nummer: 6840-45-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR