5-(Chloromethyl)-2-(4-methoxyphenyl)pyridine
Lieferant: Apollo Scientific
|
Achtung
|
OR14197-1G
APOSOR14197-1GEA
88.6
EUR
APOSOR14197-1G
5-(Chloromethyl)-2-(4-methoxyphenyl)pyridine
5-(Chloromethyl)-2-(4-methoxyphenyl)pyridine
Formel:
C₁₃H₁₂ClNO Schmelzpunkt: 46…49 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06200955 CAS-Nummer: 886361-72-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...