1-(4-Bromphenyl)piperidin 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR1420-250MG
APOSOR1420-250MGEA
21.9
EUR
APOSOR1420-250MG
1-(4-Bromphenyl)piperidin 98%
1-(4-Bromphenyl)piperidin
Formel:
C₁₁H₁₄BrN Molecular Weight: 240,14 g/mol Schmelzpunkt: 74,5…76.1 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04112533 CAS-Nummer: 22148-20-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...