Ethyl 2-chloro-2-(2-(p-tolyl)hydrazono)acetate

Lieferant: Apollo Scientific
Achtung

OR14210-1G
APOSOR14210-1GEA 0 EUR
APOSOR14210-1G
Ethyl 2-chloro-2-(2-(p-tolyl)hydrazono)acetate
Ethyl 2-chloro-2-(2-(p-tolyl)hydrazono)acetate
Formel: C₁₁H₁₃ClN₂O₂
Schmelzpunkt: 103…105 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00266044
CAS-Nummer: 27171-88-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR