[3-(4-Methoxyphenyl)-5-isoxazolyl]methanol

Lieferant: Apollo Scientific
Achtung

OR14216-250MG
APOSOR14216-250MGEA 21.9 EUR
APOSOR14216-250MG
[3-(4-Methoxyphenyl)-5-isoxazolyl]methanol
[3-(4-Methoxyphenyl)-5-isoxazolyl]methanol
Formel: C₁₁H₁₁NO₃
Schmelzpunkt: 97…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04124287
CAS-Nummer: 206055-86-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR