5-Amino-2-(4-methoxyphenyl)pyridin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-(4-Methoxyphenyl)pyridin-3-amine
OR14219-250MG
APOSOR14219-250MGEA
75.4
EUR
APOSOR14219-250MG
5-Amino-2-(4-methoxyphenyl)pyridin
5-Amino-2-(4-methoxyphenyl)pyridin
Formel:
C₁₂H₁₂N₂O Schmelzpunkt: 110…112 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05864840 CAS-Nummer: 52057-98-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...