4-(4-Bromophenyl)-2-methyl-1,3-thiazole 97+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-4-(2-methyl-1,3-thiazol-4-yl)benzene

OR1423-1G
APOSOR1423-1GEA 79.9 EUR
APOSOR1423-1G
4-(4-Bromophenyl)-2-methyl-1,3-thiazole 97+%
4-(4-Bromophenyl)-2-methyl-1,3-thiazole
Formel: C₁₀H₈BrNS
Molecular Weight: 254,146 g/mol
Schmelzpunkt: 130…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00098207
CAS-Nummer: 66047-74-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR