4-Acetyl-8-benzyl-1-thia-4,8-diazaspiro[4.5]decane-3-carboxylic acid hydrochloride

Lieferant: Apollo Scientific
Achtung

OR14236-1G
APOSOR14236-1GEA 50.9 EUR
APOSOR14236-1G
4-Acetyl-8-benzyl-1-thia-4,8-diazaspiro[4.5]decane-3-carboxylic acid hydrochloride
4-Acetyl-8-benzyl-1-thia-4,8-diazaspiro[4.5]decane-3-carboxylic acid hydrochloride
Formel: C₁₇H₂₃ClN₂O₃S
Schmelzpunkt: 251…253 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06795478
CAS-Nummer: 1030253-99-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR