2-Amino-6-methyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carbonitrile

Lieferant: Apollo Scientific
Achtung

OR14237-5G
APOSOR14237-5GEA 357 EUR
APOSOR14237-5G
2-Amino-6-methyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carbonitrile
2-Amino-6-methyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carbonitrile
Formel: C₉H₁₁N₃S
Schmelzpunkt: 193…197 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00712387
CAS-Nummer: 37578-06-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR