4-Brombenzamidoxim
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-N-hydroxybenzimidamide
OR14239-1G
APOSOR14239-1GEA
21.9
EUR
APOSOR14239-1G
4-Brombenzamidoxim
4-Brombenzamidoxim
Formel:
C₇H₇BrN₂O Molecular Weight: 215.05 g/mol Schmelzpunkt: 142…144 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00019951 CAS-Nummer: 19227-14-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...