tert-Butyl-3-(4-nitrophenoxy)tetrahydro-1(2H)-pyridinecarboxylate 97%

Lieferant: Apollo Scientific
Achtung

OR14243-250MG
APOSOR14243-250MGEA 145 EUR
APOSOR14243-250MG
tert-Butyl-3-(4-nitrophenoxy)tetrahydro-1(2H)-pyridinecarboxylate 97%
tert-Butyl-3-(4-nitrophenoxy)tetrahydro-1(2H)-pyridinecarboxylate
Formel: C₁₆H₂₂N₂O₅
Molecular Weight: 322,36 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05865146
CAS-Nummer: 690632-16-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR